Learn More
Cayman Chemical ANTIBODY UT-11 5mg
SDP
Catalog No. 502919478
Encompass_Preferred
An mPGES-1 inhibitor; inhibits PGE2 production in both SK-N-AS human and BV-2 mouse cells (IC50s = 0.1 and 2 µM, respectively); reduces LPS-induced increases in a variety of cytokines, including CCL2, IL-6, and TNF-α, in the mouse hippocampus, but not kidney, without affecting the mRNA expression of Cox2 at 10 mg/kg, i.p.
Catalog No. 50-291-9478 Supplier Cayman Chemical Supplier No. 385805 MG
An mPGES-1 inhibitor; inhibits PGE2 production in both SK-N-AS human and BV-2 mouse cells (IC50s = 0.1 and 2 µM, respectively); reduces LPS-induced increases in a variety of cytokines, including CCL2, IL-6, and TNF-α, in the mouse hippocampus, but not kidney, without affecting the mRNA expression of Cox2 at 10 mg/kg, i.p.