Learn More
Cayman Chemical Vapendavir, Purity: ≥95%, CAS: 439085-51-5, 5 mg
SDP
Catalog No. 502990856
Encompass_Preferred
A capsid-binding antiviral agent; inhibits the cytopathic effects of HRV2 and HRV14 in HeLa Ohio cells (EC50s = 1 and 5 ng/ml, respectively), as well as in a panel of 39 HRV clinical isolates (median EC50 = 7.3 ng/ml); inhibits replication in a panel of 21 EV71 isolates (average EC50 = 0.7 µM)
Catalog No. 50-299-0856 Supplier Cayman Chemical Supplier No. 305465 MG
A capsid-binding antiviral agent; inhibits the cytopathic effects of HRV2 and HRV14 in HeLa Ohio cells (EC50s = 1 and 5 ng/ml, respectively), as well as in a panel of 39 HRV clinical isolates (median EC50 = 7.3 ng/ml); inhibits replication in a panel of 21 EV71 isolates (average EC50 = 0.7 µM)