Learn More
Cayman Chemical VcMMAE, Purity: ≥98%, CAS: 646502-53-6, 1 mg
SDP
Catalog No. 502909961
Encompass_Preferred
A drug-linker molecule; has been used in the synthesis of ADCs
Catalog No. 50-290-9961 Supplier Cayman Chemical Supplier No. 356461 MG
A drug-linker molecule; has been used in the synthesis of ADCs