Learn More
Cayman Chemical ANTIBODY Vedaprofen 500ug
SDP
Supplier:  Cayman Chemical 15960500 UG
Encompass_Preferred
An NSAID commonly used in veterinary medicine to combat pain, inflammation, and fever; blocks the activity of the E. coli DNA polymerase III β subunit, preventing DNA replication and repair
Catalog No. 50-296-9035