Learn More
Cayman Chemical VER-49009, Purity: ≥98%, CAS: 558640-51-0, 10 mg
SDP
Catalog No. 502953161
Encompass_Preferred
An Hsp90 inhibitor with an IC50 value of 47 nM; induces the expression of HSP27 and Hsp72 while reducing the client proteins c-Raf, B-Raf, survivin, and PRMT5, causing cell cycle arrest and apoptosis
Catalog No. 50-295-3161 Supplier Cayman Chemical Supplier No. 1313110 MG
An Hsp90 inhibitor with an IC50 value of 47 nM; induces the expression of HSP27 and Hsp72 while reducing the client proteins c-Raf, B-Raf, survivin, and PRMT5, causing cell cycle arrest and apoptosis