Learn More
Cayman Chemical ANTIBODY VER-50589 500ug
SDP
Catalog No. 502955716
Encompass_Preferred
An isoxazole compound that inhibits Hsp90 with an IC50 value of 21 nM; produces a mean cellular antiproliferative GI50 value of 78 nM when tested against a human cancer cell line panel
Catalog No. 50-295-5716 Supplier Cayman Chemical Supplier No. 13132500 UG
An isoxazole compound that inhibits Hsp90 with an IC50 value of 21 nM; produces a mean cellular antiproliferative GI50 value of 78 nM when tested against a human cancer cell line panel