Learn More
Cayman Chemical ANTIBODY VH 032 50mg
SDP
Catalog No. 502944655
Encompass_Preferred
A building block in the synthesis of PROTACs
Catalog No. 50-294-4655 Supplier Cayman Chemical Supplier No. 3998250 MG
A building block in the synthesis of PROTACs