Learn More
Cayman Chemical Vilanterol, Purity: ≥98%, CAS: 503068-34-6, 5 mg
SDP
Catalog No. 502980711
Encompass_Preferred
A β2-AR agonist; selectively induces cAMP accumulation in CHO cells expressing the β2-AR over the β1- and β3-ARs (EC50s = 0.4, 398, and 794 nM for the human receptors, respectively); inhibits contractions induced by electrical stimulation in isolated superfused guinea pig trachea strips; inhibits histamine-induced bronchospasms in guinea pigs when administered via nebulization (EC50 = 30 µM)
Catalog No. 50-298-0711 Supplier Cayman Chemical Supplier No. 207025 MG
A β2-AR agonist; selectively induces cAMP accumulation in CHO cells expressing the β2-AR over the β1- and β3-ARs (EC50s = 0.4, 398, and 794 nM for the human receptors, respectively); inhibits contractions induced by electrical stimulation in isolated superfused guinea pig trachea strips; inhibits histamine-induced bronchospasms in guinea pigs when administered via nebulization (EC50 = 30 µM)