Learn More
Cayman Chemical Vitamin K4, Purity: ≥98%, CAS: 573-20-6, 100 g
SDP
Catalog No. 502947314
Encompass_Preferred
A synthetic form of vitamin K; inhibits proliferation of PC3 cells (IC50 = 20.94 µM); induces cell cycle arrest at the S phase and mitochondrial apoptosis in PC3 cells at 20 and 40 µM; inhibits canonical and non-canonical activation of the NLRP3 inflammasome in mouse BMDMs at 5 µM; decreases the production of IL-1β and peritoneal neutrophil infiltration in a mouse model of monosodium urate-induced NLRP3-dependent peritonitis at 4 mg/kg
Catalog No. 50-294-7314 Supplier Cayman Chemical Supplier No. 33970100 G
A synthetic form of vitamin K; inhibits proliferation of PC3 cells (IC50 = 20.94 µM); induces cell cycle arrest at the S phase and mitochondrial apoptosis in PC3 cells at 20 and 40 µM; inhibits canonical and non-canonical activation of the NLRP3 inflammasome in mouse BMDMs at 5 µM; decreases the production of IL-1β and peritoneal neutrophil infiltration in a mouse model of monosodium urate-induced NLRP3-dependent peritonitis at 4 mg/kg