Learn More
Cayman Chemical VL285, Purity: ≥98%, CAS: 1448188-57-5, 5 mg
SDP
Catalog No. 502978372
Encompass_Preferred
A starting material that has been used in the synthesis of PROTACs
Catalog No. 50-297-8372 Supplier Cayman Chemical Supplier No. 415385 MG
A starting material that has been used in the synthesis of PROTACs