Learn More
Cayman Chemical VP-14637, Purity: ≥98%, CAS: 235106-62-4, 5 mg
SDP
Catalog No. 502968493
Encompass_Preferred
An RSV inhibitor that demonstrates antiviral activity with an EC50 value of 1.4 nM; interferes directly with RSV fusion (EC50 = 5.4 nM), binding to the viral F protein
Catalog No. 50-296-8493 Supplier Cayman Chemical Supplier No. 178055 MG
An RSV inhibitor that demonstrates antiviral activity with an EC50 value of 1.4 nM; interferes directly with RSV fusion (EC50 = 5.4 nM), binding to the viral F protein