Learn More
Cayman Chemical VU0361737, Purity: ≥98%, CAS: 1161205-04-4, 10 mg
SDP
Catalog No. 502970499
Encompass_Preferred
A selective PAM of mGluR4, displaying EC50 values of 110 and 240 nM for rat and human receptors, respectively; passes the blood-brain barrier to act centrally in rats
Catalog No. 50-297-0499 Supplier Cayman Chemical Supplier No. 1629510 MG
A selective PAM of mGluR4, displaying EC50 values of 110 and 240 nM for rat and human receptors, respectively; passes the blood-brain barrier to act centrally in rats