Learn More
Cayman Chemical WAY-262611, Purity: ≥98%, CAS: 1123231-07-1, 1 mg
SDP
Catalog No. 502980039
Encompass_Preferred
An inhibitor of Dkk1 that activates the Wnt signaling pathway, stimulating β-catenin/TCF-dependent transcription with an EC50 value of 0.63 µM; has good pharmacokinetic properties and increases trabecular bone formation in ovariectomized rats following oral administration
Catalog No. 50-298-0039 Supplier Cayman Chemical Supplier No. 177041 MG
An inhibitor of Dkk1 that activates the Wnt signaling pathway, stimulating β-catenin/TCF-dependent transcription with an EC50 value of 0.63 µM; has good pharmacokinetic properties and increases trabecular bone formation in ovariectomized rats following oral administration