Learn More
Cayman Chemical WAY-316606 (hydrochloride), Purity: ≥98%, CAS: 1781835-02-6, 5 mg
SDP
Catalog No. 502933387
Encompass_Preferred
An sFRP1 inhibitor (IC50 = 0.5 μM); increases Wnt signaling in U2OS osteosarcoma cells (EC50 = 0.65 μM); increases total bone area in a murine calvarial organ culture assay at >0.1 nM
Catalog No. 50-293-3387 Supplier Cayman Chemical Supplier No. 263245 MG
An sFRP1 inhibitor (IC50 = 0.5 μM); increases Wnt signaling in U2OS osteosarcoma cells (EC50 = 0.65 μM); increases total bone area in a murine calvarial organ culture assay at >0.1 nM