Learn More
Cayman Chemical WHI-P258, Purity: ≥98%, CAS: 21561-09-1, 25 mg
SDP
Catalog No. 502986889
Encompass_Preferred
A negative control for JAK3 inhibition; molecular modeling estimated a Ki value of 72 µM; inactive at JAK3 (IC50 = >300 µM)
Catalog No. 50-298-6889 Supplier Cayman Chemical Supplier No. 2345325 MG
A negative control for JAK3 inhibition; molecular modeling estimated a Ki value of 72 µM; inactive at JAK3 (IC50 = >300 µM)