Learn More
Cayman Chemical ANTIBODY WYE-23 500ug
SDP
Catalog No. 502980989
Encompass_Preferred
An inhibitor of mTOR (IC50 = 0.45 nM); selective for mTOR over PI3Kα (IC50 = 661 nM); inhibits cell growth in LNCaP cells (IC50 = 42 nM)
Catalog No. 50-298-0989 Supplier Cayman Chemical Supplier No. 21199500 UG
An inhibitor of mTOR (IC50 = 0.45 nM); selective for mTOR over PI3Kα (IC50 = 661 nM); inhibits cell growth in LNCaP cells (IC50 = 42 nM)