Learn More
Cayman Chemical WYneN, Purity: ≥95%, CAS: 3052492-38-0, 50 mg
SDP
Catalog No. 502978454
Encompass_Preferred
A chemoselective probe for protein sulfenylation; selective for sulfenylated proteins over DNTB and 4-DPS; binds to sulfenylated proteins in live cells and can then be conjugated to azide-containing fluorescent probes for detection
Catalog No. 50-297-8454 Supplier Cayman Chemical Supplier No. 4201050 MG
A chemoselective probe for protein sulfenylation; selective for sulfenylated proteins over DNTB and 4-DPS; binds to sulfenylated proteins in live cells and can then be conjugated to azide-containing fluorescent probes for detection