Learn More
Cayman Chemical WZ4002, Purity: ≥98%, CAS: 1213269-23-8, 100 mg
SDP
Catalog No. 502970441
Encompass_Preferred
An irreversible EGFR tyrosine kinase inhibitor that blocks ATP-dependent autophosphorylation of EGFR carrying the T790M mutation as well as an L858R mutation (IC50 = 8 nM); effective in mouse models of lung cancer driven by EGFRT790M
Catalog No. 50-297-0441 Supplier Cayman Chemical Supplier No. 16297100 MG
An irreversible EGFR tyrosine kinase inhibitor that blocks ATP-dependent autophosphorylation of EGFR carrying the T790M mutation as well as an L858R mutation (IC50 = 8 nM); effective in mouse models of lung cancer driven by EGFRT790M