Learn More
Cayman Chemical XCT790, Purity: ≥98%, CAS: 725247-18-7, 10 mg
SDP
Catalog No. 502969502
Encompass_Preferred
A selective inverse agonist of ERRα (IC50 = ~300-500 nM); blocks ERRα/PGC-1α-dependent signaling, suppressing the expression of MAO A and B; induces proteasomal degradation of ERRα and potentiates the degradation of ERα by fulvestrant
Catalog No. 50-296-9502 Supplier Cayman Chemical Supplier No. 1603510 MG
A selective inverse agonist of ERRα (IC50 = ~300-500 nM); blocks ERRα/PGC-1α-dependent signaling, suppressing the expression of MAO A and B; induces proteasomal degradation of ERRα and potentiates the degradation of ERα by fulvestrant