Learn More
Cayman Chemical XL335, Purity: ≥98%, CAS: 629664-81-9, 5 mg
SDP
Catalog No. 502993850
Encompass_Preferred
A potent and selective FXR agonist (EC50 = 4 nM in CV-1 cells transfected with human FXR); selective for FXR over a panel of 15 nuclear receptors at concentrations up to 10 μM; induces a 20-, 13-, and 2-fold increase in mRNA expression for IBABP, BSEP, and SHP, respectively, at a concentration of 1 μM; reduces plasma cholesterol and triglycerides and the area of pre-atherosclerotic lesions in LDLR-/- mice fed a Western diet; decreases hepatic fibrosis in a mouse model of non-alcoholic steatohepatitis
Catalog No. 50-299-3850 Supplier Cayman Chemical Supplier No. 233505 MG
A potent and selective FXR agonist (EC50 = 4 nM in CV-1 cells transfected with human FXR); selective for FXR over a panel of 15 nuclear receptors at concentrations up to 10 μM; induces a 20-, 13-, and 2-fold increase in mRNA expression for IBABP, BSEP, and SHP, respectively, at a concentration of 1 μM; reduces plasma cholesterol and triglycerides and the area of pre-atherosclerotic lesions in LDLR-/- mice fed a Western diet; decreases hepatic fibrosis in a mouse model of non-alcoholic steatohepatitis