Learn More
Cayman Chemical Xanthine, Purity: ≥98%, CAS: 69-89-6, 10 g
SDP
Catalog No. 502990972
Encompass_Preferred
A purine base and intermediate in uric acid biosynthesis; been found in a variety of commercial foodstuffs; urinary levels are decreased in patients with primary gout; has been used in the synthesis of xanthine derivatives that have anticancer or anti-inflammatory activities in vitro
Catalog No. 50-299-0972 Supplier Cayman Chemical Supplier No. 3149910 G
A purine base and intermediate in uric acid biosynthesis; been found in a variety of commercial foodstuffs; urinary levels are decreased in patients with primary gout; has been used in the synthesis of xanthine derivatives that have anticancer or anti-inflammatory activities in vitro