Learn More
Cayman Chemical Y-26763, Purity: ≥98%, CAS: 127408-31-5, 1 mg
SDP
Catalog No. 502994027
Encompass_Preferred
An activator of KATP channels; increases the frequency of KATP channel openings in the presence of ATP and activates KATP channel currents in isolated human β-cells at 200 μM; inhibits increases in intracellular calcium induced by histamine or norepinephrine in isolated rabbit mesenteric artery smooth muscle in a concentration-dependent manner; induces relaxation in isolated precontracted rat aortic rings (IC50 = 0.027 μM); decreases mean blood pressure in spontaneously hypertensive rats when administered intravenously at 0.03 mg/kg
Catalog No. 50-299-4027 Supplier Cayman Chemical Supplier No. 214591 MG
An activator of KATP channels; increases the frequency of KATP channel openings in the presence of ATP and activates KATP channel currents in isolated human β-cells at 200 μM; inhibits increases in intracellular calcium induced by histamine or norepinephrine in isolated rabbit mesenteric artery smooth muscle in a concentration-dependent manner; induces relaxation in isolated precontracted rat aortic rings (IC50 = 0.027 μM); decreases mean blood pressure in spontaneously hypertensive rats when administered intravenously at 0.03 mg/kg