Learn More
Cayman Chemical YC-1, Purity: ≥99%, CAS: 170632-47-0, 1 mg
SDP
Catalog No. 502896655
Encompass_Preferred
YC-1 is an NO-independent activator of soluble guanylyl cyclase. It inhibits collagen (10 µg/ml) and thrombin (0.1 U/ml) induced platelet aggregation with IC50 values of 11.7-14.6 and 57.3-59.3 µM, respectively.{4218,4219} In the absence of NO or CO, 200 µM YC-1 increases the activity of purified soluble guanylyl cyclase 12-fold, with an ED50 value of 20 µM.{4220} At a concentration of 200 µM, YC-1 potentiates the maximal activity of soluble guanylyl cyclase in the presence of NO or CO by 40% and >3,000%, respectively.
Catalog No. 50-289-6655 Supplier Cayman Chemical Supplier No. 815601 MG
YC-1 is an NO-independent activator of soluble guanylyl cyclase. It inhibits collagen (10 µg/ml) and thrombin (0.1 U/ml) induced platelet aggregation with IC50 values of 11.7-14.6 and 57.3-59.3 µM, respectively.{4218,4219} In the absence of NO or CO, 200 µM YC-1 increases the activity of purified soluble guanylyl cyclase 12-fold, with an ED50 value of 20 µM.{4220} At a concentration of 200 µM, YC-1 potentiates the maximal activity of soluble guanylyl cyclase in the presence of NO or CO by 40% and >3,000%, respectively.