Learn More
Cayman Chemical YK-009, Purity: ≥95%, CAS: 2761458-86-8, 10 mg
SDP
Catalog No. 502916405
Encompass_Preferred
An ionizable cationic lipid (pKa = 6.512); administration of SARS-CoV-2 Omicron variant mRNA in YK009-containing LNPs induces SARS-CoV-2 RBD-binding IgG in mice, with a boost in antigen-specific IgG titers 10 days after a second dose
Catalog No. 50-291-6405 Supplier Cayman Chemical Supplier No. 3828210 MG
An ionizable cationic lipid (pKa = 6.512); administration of SARS-CoV-2 Omicron variant mRNA in YK009-containing LNPs induces SARS-CoV-2 RBD-binding IgG in mice, with a boost in antigen-specific IgG titers 10 days after a second dose