Learn More
Cayman Chemical Z-FA-FMK, Purity: ≥98%, CAS: 197855-65-5, 5 mg
SDP
Catalog No. 502971779
Encompass_Preferred
An irreversible inhibitor of cysteine proteases, including cathepsins B, L, and S, cruzain, and papain; inhibits effector caspases-2, -3, -6, and -7 (IC50 = 6-32 µM) without affecting the initiator caspases-8 and -10; can be used both in cells and in vivo
Catalog No. 50-297-1779 Supplier Cayman Chemical Supplier No. 144625 MG
An irreversible inhibitor of cysteine proteases, including cathepsins B, L, and S, cruzain, and papain; inhibits effector caspases-2, -3, -6, and -7 (IC50 = 6-32 µM) without affecting the initiator caspases-8 and -10; can be used both in cells and in vivo