Learn More
Cayman Chemical Z-FF-FMK, Purity: ≥95%, CAS: 105608-85-3, 50 mg
SDP
Catalog No. 502944148
Encompass_Preferred
An inhibitor of cathepsin B (Ki = 2.7 nM); inhibits cell division, cell cycle progression, DNA replication, and chromosome decondensation in isolated T. niger fertilized embryos at 100 µM; prevents Aβ40-induced increases in cathepsin L activity, as well as prevents Aβ40-induced apoptosis, DNA fragmentation, and increases in cleaved PARP levels, in primary rat cortical neurons at 10 µM; inhibits quinolinic acid-induced nuclear translocation of NF-κB, as well as inhibits quinolinic acid-induced increases in phosphorylated IκBα and IKKα levels, in the unilateral striatum in rats at 5 nmol/animal
Catalog No. 50-294-4148 Supplier Cayman Chemical Supplier No. 4102650 MG
An inhibitor of cathepsin B (Ki = 2.7 nM); inhibits cell division, cell cycle progression, DNA replication, and chromosome decondensation in isolated T. niger fertilized embryos at 100 µM; prevents Aβ40-induced increases in cathepsin L activity, as well as prevents Aβ40-induced apoptosis, DNA fragmentation, and increases in cleaved PARP levels, in primary rat cortical neurons at 10 µM; inhibits quinolinic acid-induced nuclear translocation of NF-κB, as well as inhibits quinolinic acid-induced increases in phosphorylated IκBα and IKKα levels, in the unilateral striatum in rats at 5 nmol/animal