Learn More
Cayman Chemical Z-Lys-OH, Purity: ≥98%, CAS: 2212-75-1, 5 g
SDP
Catalog No. 502992559
Encompass_Preferred
An amino acid building block; has been used in the synthesis of peptidomimetics and polypeptide copolymers
Catalog No. 50-299-2559 Supplier Cayman Chemical Supplier No. 304605 G
An amino acid building block; has been used in the synthesis of peptidomimetics and polypeptide copolymers