Learn More
Cayman Chemical ANTIBODY ZD 7288 50mg
SDP
Catalog No. 502972691
Encompass_Preferred
A potent blocker of HCN channels that inhibits HCN4 more potently than HCN1 (IC50s = 32 and 158 nM, respectively); modulates the sinoatrial node and slows heart rate; reduces neuropathic pain in several animal models
Catalog No. 50-297-2691 Supplier Cayman Chemical Supplier No. 1522850 MG
A potent blocker of HCN channels that inhibits HCN4 more potently than HCN1 (IC50s = 32 and 158 nM, respectively); modulates the sinoatrial node and slows heart rate; reduces neuropathic pain in several animal models