Learn More
Cayman Chemical ZK 824859, Purity: ≥98%, CAS: 2254001-81-3, 50 mg
SDP
Catalog No. 502915872
Encompass_Preferred
An inhibitor of uPA (IC50s = 79 and 410 nM for the human and mouse proteins, respectively); selective for human uPA over human tPA and plasmin (IC50s = 1,580 and 1,330 nM, respectively) but not selective for mouse uPA over mouse tPA and plasmin (IC50s = 910 and 1,600 nM, respectively); reduces the incidence of limb weakness and paralysis in mice in an EAE model at 25 or 50 mg/kg per day
Catalog No. 50-291-5872 Supplier Cayman Chemical Supplier No. 3778950 MG
An inhibitor of uPA (IC50s = 79 and 410 nM for the human and mouse proteins, respectively); selective for human uPA over human tPA and plasmin (IC50s = 1,580 and 1,330 nM, respectively) but not selective for mouse uPA over mouse tPA and plasmin (IC50s = 910 and 1,600 nM, respectively); reduces the incidence of limb weakness and paralysis in mice in an EAE model at 25 or 50 mg/kg per day