Learn More
Cayman Chemical ZLN024, Purity: ≥98%, CAS: 723249-01-2, 5 mg
SDP
Catalog No. 502986120
Encompass_Preferred
An allosteric activator of AMPK heterotrimers (EC50s = 0.42 and 0.95 µM for α1β1γ1 and α2β1γ1, respectively); reduces liver triacylglycerol and total cholesterol content and improves glucose tolerance in db/db mice
Catalog No. 50-298-6120 Supplier Cayman Chemical Supplier No. 216445 MG
An allosteric activator of AMPK heterotrimers (EC50s = 0.42 and 0.95 µM for α1β1γ1 and α2β1γ1, respectively); reduces liver triacylglycerol and total cholesterol content and improves glucose tolerance in db/db mice