Learn More
Cayman Chemical Zingerone, Purity: ≥98%, CAS: 122-48-5, 25 g
SDP
Catalog No. 503015796
Encompass_Preferred
A phenolic ketone; scavenges DPPH radicals in a cell-free assay (EC50 = 41.6 μmol); prevents myocardial expression of TNF-α, IL-1β, and IL-6 in rats following isoproterenol-induced myocardial infarction at 6 mg/kg per day; decreases 1,2-dimethylhydrazine-induced tumor incidence by 67% and aberrant crypt foci formation by 68.85% in rats at 20 mg/kg per day; decreases E. coli heat-labile enterotoxin-induced fluid accumulation in the ileal loop of mice at 10 mM
Catalog No. 50-301-5796 Supplier Cayman Chemical Supplier No. 2490925 G
A phenolic ketone; scavenges DPPH radicals in a cell-free assay (EC50 = 41.6 μmol); prevents myocardial expression of TNF-α, IL-1β, and IL-6 in rats following isoproterenol-induced myocardial infarction at 6 mg/kg per day; decreases 1,2-dimethylhydrazine-induced tumor incidence by 67% and aberrant crypt foci formation by 68.85% in rats at 20 mg/kg per day; decreases E. coli heat-labile enterotoxin-induced fluid accumulation in the ileal loop of mice at 10 mM