Learn More
Cayman Chemical ZZL-7, Purity: ≥95%, CAS: 99141-91-0, 50 mg
SDP
Catalog No. 502918978
Encompass_Preferred
A SERT-nNOS protein-protein interaction inhibitor; increases in vivo serotonergic neuron firing diminished by decreased SERT levels in SERT-Cre transgenic mice, which have reduced SERT levels, at 10 mg/kg; reduces immobility time in the tail suspension test and forced swim test in mice at 20 and 40 mg/kg
Catalog No. 50-291-8978 Supplier Cayman Chemical Supplier No. 3864550 MG
A SERT-nNOS protein-protein interaction inhibitor; increases in vivo serotonergic neuron firing diminished by decreased SERT levels in SERT-Cre transgenic mice, which have reduced SERT levels, at 10 mg/kg; reduces immobility time in the tail suspension test and forced swim test in mice at 20 and 40 mg/kg