Learn More
Cayman Chemical ANTIBODYR-ErSO 10mg
SDP
Catalog No. 502910396
Encompass_Preferred
An activator of the anticipatory unfolded protein response; inhibits protein synthesis and induces intracellular ATP depletion in T47D cells at 1 µM ; increases levels of phosphorylated EIF2AK3, eIF2α, and AMPK in T47D cells at 1 µM; cytotoxic to T47D, TYS, BT474, and ZR-75-1 ERα-positive breast cancer cells (IC50s = 16.4, 11.1, 26.9, and 42.8 nM, respectively); induces tumor regression in an MCF-7 mouse xenograft model at 40 mg/kg; induces tumor regression in an ST941/HI PDX mouse model
Catalog No. 50-291-0396 Supplier Cayman Chemical Supplier No. 3568410 MG
An activator of the anticipatory unfolded protein response; inhibits protein synthesis and induces intracellular ATP depletion in T47D cells at 1 µM ; increases levels of phosphorylated EIF2AK3, eIF2α, and AMPK in T47D cells at 1 µM; cytotoxic to T47D, TYS, BT474, and ZR-75-1 ERα-positive breast cancer cells (IC50s = 16.4, 11.1, 26.9, and 42.8 nM, respectively); induces tumor regression in an MCF-7 mouse xenograft model at 40 mg/kg; induces tumor regression in an ST941/HI PDX mouse model