Learn More
Cayman Chemical ANTIBODYR-RP 6306 10mg
SDP
Catalog No. 502942857
Encompass_Preferred
A Myt1 kinase inhibitor (IC50 = 1,360 nM)
Catalog No. 50-294-2857 Supplier Cayman Chemical Supplier No. 4173810 MG
A Myt1 kinase inhibitor (IC50 = 1,360 nM)