Learn More
Cayman Chemical ANTIBODYR-RP 6306 1mg
SDP
Catalog No. 502943640
Encompass_Preferred
A Myt1 kinase inhibitor (IC50 = 1,360 nM)
Catalog No. 50-294-3640 Supplier Cayman Chemical Supplier No. 417381 MG
A Myt1 kinase inhibitor (IC50 = 1,360 nM)