Learn More
Cayman Chemical (S)-MCPG, Purity: ≥98%, CAS: 150145-89-4, 10 mg
SDP
Catalog No. 502994294
Encompass_Preferred
An mGluR antagonist; the active component of (±)-MCPG; inhibits ACPD-induced intracellular calcium increases in CHO cells expressing mGluR1α (IC50 = 120 µM); has agonist activity at mGluR2, inducing long-term depression in rat hippocampal dentate gyrus, an effect that can be blocked by an mGluR2 antagonist; blocks long-term potentiation in the CA1 region
Catalog No. 50-299-4294 Supplier Cayman Chemical Supplier No. 2145010 MG
An mGluR antagonist; the active component of (±)-MCPG; inhibits ACPD-induced intracellular calcium increases in CHO cells expressing mGluR1α (IC50 = 120 µM); has agonist activity at mGluR2, inducing long-term depression in rat hippocampal dentate gyrus, an effect that can be blocked by an mGluR2 antagonist; blocks long-term potentiation in the CA1 region