Learn More
Medchemexpress LLC Anticaries agent-1 | 198649-97-7 | ≥97.0% | 256.25 | 250 MG
SDP
Catalog No. 5000403110
Encompass_Preferred
Anticaries agent-1 (Compound 21b) is an anticaries agent. It inhibits biofilm formation (IC50: 77 μM) and *S. mutans* growth, making it suitable for research on dental health.
  • Inhibits biofilm formation (IC50: 77 μM)
  • Inhibits *S. mutans* growth
Catalog No. 50-004-03110 Supplier Medchemexpress LLC Supplier No. 250MGHY401757
Anticaries agent-1 (Compound 21b) is an anticaries agent. It inhibits biofilm formation (IC50: 77 μM) and *S. mutans* growth, making it suitable for research on dental health.
  • Inhibits biofilm formation (IC50: 77 μM)
  • Inhibits *S. mutans* growth