Learn More
Vector Laboratories Anti-CD52 [Campath-1H]
SDP
Supplier:  Vector Laboratories AB0012410.0
Encompass
Human CD52 antigen, also known as CAMPATH-1. CD52 is expressed at high density by lymphocytes, monocytes, eosinophils, thymocytes and macrophages. It is expressed by most lymphoid derived malignancies, although expression on myeloma cells is variable.
Catalog No. NC1669228