Learn More
Medchemexpress LLC Anticonvulsant agent 2 | 75220-84-7 | C21H17ClN2 | 1 ML
SDP
Catalog No. 5000376693
Encompass_Preferred
Anticonvulsant agent 2 is a potent and orally active anticonvulsant agent that exhibits antiepileptic activity. It is for research use only and not sold to patients. In vivo studies showed that Anticonvulsant agent 2 decreased locomotor activity in rats with an inhibition rate of 21.01% and demonstrated antiepileptic activity with an inhibition rate of 24.45% in rats.
  • Potent and orally active anticonvulsant agent
  • Exhibits antiepileptic activity
  • Decreases locomotor activity in rats
  • Demonstrates antiepileptic activity in rats
  • For research use only
Catalog No. 50-003-76693 Supplier Medchemexpress LLC Supplier No. HY15443610MM/1ML
Anticonvulsant agent 2 is a potent and orally active anticonvulsant agent that exhibits antiepileptic activity. It is for research use only and not sold to patients. In vivo studies showed that Anticonvulsant agent 2 decreased locomotor activity in rats with an inhibition rate of 21.01% and demonstrated antiepileptic activity with an inhibition rate of 24.45% in rats.
  • Potent and orally active anticonvulsant agent
  • Exhibits antiepileptic activity
  • Decreases locomotor activity in rats
  • Demonstrates antiepileptic activity in rats
  • For research use only