Learn More
Apexbio Technology LLC DiscoveryProbe™ Anti-diabetic Compound Library, 250uL/well(10 mM solution),96 Well Racks With Screw Cap
SDP
Catalog No. 501905632
Encompass_Preferred
A unique collection of 110 anti-diabetic small molecule compounds for diabetes research. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5632 Supplier Apexbio Technology LLC Supplier No. L1046.25
A unique collection of 110 anti-diabetic small molecule compounds for diabetes research. Other sizes are available. Please inqury us for quote.