Learn More
Sigma Aldrich Fine Chemicals Biosciences Anti-Digoxigenin-AP, Fab fragments, 200 uL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11093274910
Encompass
Digoxigenin is a hapten, useful in labeling and detection of nucleic acids. This product contains Fab fragments from polyclonal anti-digoxigenin antibodies, conjugated to alkaline phosphatase. Anti-Digoxigenin-AP, Fab fragments are useful for the detection of digoxigenin-labeled compounds.
Catalog No. NC0997769