Learn More
Med Vet International Antidote 3 Rattler AntIVenin, Crotalidae Polyvalent, 50mL, Each
Catalog No. 502827980
Encompass_Preferred
Antidote 3 Rattler Antivenin, Crotalidae Polyvalent, Equine Origin is shown to be effective against envenomation in canine, equine, and feline due to North Crotalidae. Crotalidae refers to cottonmouths/water moccasins, copperheads and rattlesnakes.
Catalog No. 50-282-7980 Supplier Med Vet International Supplier No. RXANTI350MLV
Antidote 3 Rattler Antivenin, Crotalidae Polyvalent, Equine Origin is shown to be effective against envenomation in canine, equine, and feline due to North Crotalidae. Crotalidae refers to cottonmouths/water moccasins, copperheads and rattlesnakes.