Learn More
Enzo Life Sciences Antifade mounting medium with DAPI (hardset) (10ml)
SDP
Catalog No. 502012649
Encompass_Preferred
Antifade mounting medium with DAPI (hardset) is an antifade reagent that causes little or no quenching. Use of the mounting medium significantly extends the photobleaching half-life of the fluorescent stain. The inclusion of DAPI allows for staining of the nucleus. Long Term Storage: -80°C. Applications: IF, IHC, Fluorescence microscopy, Fluorescent detection .
Catalog No. 50-201-2649 Supplier Enzo Life Sciences Supplier No. ENZ53003M010
Antifade mounting medium with DAPI (hardset) is an antifade reagent that causes little or no quenching. Use of the mounting medium significantly extends the photobleaching half-life of the fluorescent stain. The inclusion of DAPI allows for staining of the nucleus. Long Term Storage: -80°C. Applications: IF, IHC, Fluorescence microscopy, Fluorescent detection .