Learn More
Enzo Life Sciences Antifade mounting medium (hardset) (10ml)
SDP
Catalog No. 502012648
Encompass_Preferred
Antifade mounting medium (hardset) is an antifade reagent that causes little or no quenching. Use of the mounting medium significantly extends the photobleaching half-life of the fluorescent stain. Long Term Storage: -80°C. Applications: IF, IHC, Fluorescence microscopy, Fluorescent detection .
Catalog No. 50-201-2648 Supplier Enzo Life Sciences Supplier No. ENZ53002M010
Antifade mounting medium (hardset) is an antifade reagent that causes little or no quenching. Use of the mounting medium significantly extends the photobleaching half-life of the fluorescent stain. Long Term Storage: -80°C. Applications: IF, IHC, Fluorescence microscopy, Fluorescent detection .