Learn More
Vector Laboratories VECTASHIELD PLUS Antifade Mounting Medium with DAPI | 10mL
SDP
Catalog No. NC1848444
Encompass
VECTASHIELD PLUS Antifade Mounting Medium with DAPI is a unique, stable formula for preserving fluorescence. This medium inhibits rapid photobleaching of fluorescent proteins and fluorescent dyes.
Catalog No. NC1848444 Supplier Vector Laboratories Supplier No. H200010
VECTASHIELD PLUS Antifade Mounting Medium with DAPI is a unique, stable formula for preserving fluorescence. This medium inhibits rapid photobleaching of fluorescent proteins and fluorescent dyes.