Learn More
Vector Laboratories VECTASHIELD PLUS Antifade Mounting Medium with DAPI | 2mL
SDP
Catalog No. NC1848443
Encompass
VECTASHIELD PLUS Antifade Mounting Medium with DAPI is a unique, stable formula for preserving fluorescence. This medium inhibits rapid photobleaching of fluorescent proteins and fluorescent dyes.
Catalog No. NC1848443 Supplier Vector Laboratories Supplier No. H20002
VECTASHIELD PLUS Antifade Mounting Medium with DAPI is a unique, stable formula for preserving fluorescence. This medium inhibits rapid photobleaching of fluorescent proteins and fluorescent dyes.