Learn More
Sigma Aldrich Fine Chemicals Biosciences Antifoam A Concentrate | 25g
Catalog No. NC1813465
Encompass
Antifoam A Concentrate is an extremely effective foam suppressor[1] for aqueous and non-aqueous systems. It is 100% active silicone polymer. No emulsifiers are present. Antifoam A Concentrate is typically effective at 1-100 ppm. Antifoam A Concentrate should be diluted with 3-10 parts of propylene glycol (aqueous) or vegetable oil (nonaqueous) with slow mixing. The product will be stable in the pH range of 5 to 9. Antifoam A Concentrate can be added directly to a fermentation medium, but it is not recommended that it be pumped to a fermenter on an as-needed basis.
Catalog No. NC1813465 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A563325G
Antifoam A Concentrate is an extremely effective foam suppressor[1] for aqueous and non-aqueous systems. It is 100% active silicone polymer. No emulsifiers are present. Antifoam A Concentrate is typically effective at 1-100 ppm. Antifoam A Concentrate should be diluted with 3-10 parts of propylene glycol (aqueous) or vegetable oil (nonaqueous) with slow mixing. The product will be stable in the pH range of 5 to 9. Antifoam A Concentrate can be added directly to a fermentation medium, but it is not recommended that it be pumped to a fermenter on an as-needed basis.