Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Antifoam AF, Emulsion, Food Grade, Spectrum™ Chemical
SDP

Catalog No. 18600729 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Cubitainer
Drum
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-729 500 mL Poly Bottle
18-600-728 100 mL Poly Bottle
18-600-730 4 L Poly Bottle
18-600-732 20 L Cubitainer
18-600-731 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 18-600-729 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1303500MLPL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

A1303

Spectrum™ Chemical Antifoam AF, Emulsion, Food Grade is also known as polydimethylsiloxane and is part of a group of polymeric organosilicon compounds known as silicones. It is a chemical additive that reduces and hinders the formation of foam in industrial process liquids. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 67762-90-7

Specifications

CAS Max % 100%
Quantity 500 mL
Grade Food Grade
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.