Learn More
Sigma Aldrich Fine Chemicals Biosciences Antifoam B Emulsion, 500mL
Catalog No. NC1659900
Encompass
Antifoam eliminates excessive foaming formed during impingement, due to proteins and/or carbohydrates in the collection medium. Antifoam B emulsion is a silicone based antifoam.
Catalog No. NC1659900 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A5757500ML
Antifoam eliminates excessive foaming formed during impingement, due to proteins and/or carbohydrates in the collection medium. Antifoam B emulsion is a silicone based antifoam.