Learn More
Supelco Inc Antifoam C Emulsion, 500 mL
Catalog No. NC1836065
Encompass
A 30% aqueous emulsion of polydimethylsiloxane concentrate.
Catalog No. NC1836065 Supplier Supelco Inc Supplier No. A8011500ML
A 30% aqueous emulsion of polydimethylsiloxane concentrate.